Skip to Content
Merck
All Photos(1)

Key Documents

MABC1233

Sigma-Aldrich

Anti-Rae1 Antibody, clone 14G7.1

clone 14G7.1, from mouse

Synonym(s):

mRNA export factor, Rae1 protein homolog, mRNA-associated protein mrnp 41

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€130.00

€130.00


Estimated to ship on12 March 2025Details


Request a Bulk Order

Select a Size

Change View
25 μL
€130.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€130.00


Estimated to ship on12 March 2025Details


Request a Bulk Order

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

14G7.1, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μL

technique(s)

western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... RAE1(8480)

General description

mRNA export factor (UniProt: P78406; also known as Rae1 protein homolog, mRNA-associated protein mrnp 41) is encoded by the RAE1 (also known as MRNP41) gene (Gene ID: 8480) in human. mRNA export factor (UniProt: P78406; also known as RAE1 protein homolog, mRNA-associated protein mrnp 41) is encoded by the RAE1 (also known as MRNP41) gene (Gene ID: 8480) in human. Rae1 is a member of the superfamily of WD-repeat proteins that plays a role in mitotic bipolar spindle formation. It contains seven WD repeats. Rae1 is also reported to bind to a GLEBS-like NUP98 motif at the nuclear pore complex through multiple domains, including WD-repeats. Also, the COOH-Terminal non- WD-repeat extension of Rae1 Is shown to be essential for NUP98 binding. Rae1 plays a role in nucleocytoplasmic transport and in directly or indirectly attaching cytoplasmic mRNPs to the cytoskeleton. It is recruited from interphase nuclei to spindle microtubules during mitosis. RAE1 also interacts with NUMA1 via N-terminal end of the coiled-coil domain and this interaction promotes spindle formation in mitosis. (Ref.: Pritchard, CEJ et al. (1999). J. cell Biol. 145(2); 237-254).

Specificity

Clone 14G7.1 specifically detects human mRNA export factor (Rae1). it targets an epitope with in the C-terminal half.

Immunogen

GST/His-tagged recombinant fragment corresponding to 179 amino acids from the C-terminal half of human mRNA export factor (Rae1).

Application

Anti-Rae1, clone 14G7.1, Cat. No. MABC1233, is a mouse monoclonal antibody that detects mRNA export factor (Rae1) and has been tested for use in Western Blotting.
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Rae1 in HEK293 cell lysate.

Target description

~40 kDa observed; 40.97 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service