Skip to Content
Merck
All Photos(1)

Key Documents

MAB4201F

Sigma-Aldrich

Anti-HLA-DR Antibody, clone 169-1B5.1, FITC conjugated

clone 169-1B5.1, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing

Select a Size

100 ASSAYS
€435.00

€435.00


Estimated to ship on04 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 ASSAYS
€435.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€435.00


Estimated to ship on04 April 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

FITC conjugate

antibody product type

primary antibodies

clone

169-1B5.1, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable

isotype

IgG2b

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... HLA-DRA(3122)

Specificity

This antibody detects a monomorphic general framework determinant of HLA-DR Class II antigen. It reacts with human B-lymphocytes, monocytes, macrophages, activated T-cells, thymic epithelium, B-cell lymphomas and Langerhans cells. In addition this antibody shows broad species cross-reactivity and has been used to identify Class II antigens on bovine and feline B-cells2.

Immunogen

Human peripheral blood leukocytes.

Application

Detect HLA-DR using this Anti-HLA-DR Antibody, clone 169-1B5.1, FITC conjugated validated for use in FC.
This antibody may be used for the classification of leukemias and lymphomas, and immunophenotyping by flow cytometry.

SUGGESTED USAGE

Flow cytometry and immunofluorescence - use 10 μl direct from the vial per 100 μl of whole blood, or 1 x 10E6 peripheral blood mononuclear cells (PBMC) in 100 μl buffer.

Physical form

The antibody is supplied in 1.0ml phosphate buffered saline, pH 7.4, containing 0.2% bovine serum albumin and 0.1% sodium azide. The characteristics of each lot are tested by electrophoresis and flow cytometry.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service