Skip to Content
Merck
All Photos(1)

Key Documents

ABN467

Sigma-Aldrich

Anti-CX3CL1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Fractalkine, C-X3-C motif chemokine 1, CX3C membrane-anchored chemokine, Neurotactin, Small-inducible cytokine D1, FKN, NTT, SCYD1, A-152E5.2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€370.00

€370.00


Estimated to ship on03 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€370.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€370.00


Estimated to ship on03 March 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

species reactivity (predicted by homology)

human (immunogen homology)

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Cx3Cl1(20312)

General description

C-X3-C motif chemokine 1 (CX3CL1), also known as CX3C membrane-anchored chemokine, Neurotactin (NTT), Small-inducible cytokine D1 (SCYD1), and Fractalkine (FKN), is a CX3C chemokine known to induce adhesion and migration of leukocytes. Recent studies have shown that CX3CL1 can suppress the production of nitrous oxide, interleukin-6, and TNF-alpha in activated microglia and neuronal cells, and it may act as an intrinsic inhibitor against neurotoxicity by activated microglia. Other evidence suggests that CX3CL1 may play a role in regulating leukocyte adhesion and migration processes at the endothelium. Additionally, CX3CL1 is implied to inhibit CX3CR1, which is a co-receptor for HIV-1 and HIV-2 envelope fusion and virus infection. CX3CL1 is expressed in small intestine, colon, testis, prostate, heart, brain, lung, skeletal muscle, kidney and pancreas.

Immunogen

KLH-conjugated linear peptide corresponding to human CX3CL1.

Application

Anti-CX3CL1 Antibody is a highly specific rabbit polyclonal antibody, that targets CX3CL1 & has been tested in western blotting.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Quality

Evaluated by Western Blotting in C2C12 cell lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected CX3CL1 in 15 µg of C2C12 cell lysate.

Target description

~50 kDa observed.

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
C2C12 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service