260962
DNA-PK Inhibitor III
The DNA-PK Inhibitor III, also referenced under CAS 404009-40-1, controls the biological activity of DNA-PK. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Synonym(s):
DNA-PK Inhibitor III, IC86621, 1-(2-Hydroxy-4-morpholin-4-yl-phenyl)ethanone
About This Item
Quality Level
Assay
≥95% (HPLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
off-white
solubility
DMSO: 5 mg/mL
shipped in
ambient
storage temp.
2-8°C
InChI
1S/C12H15NO3/c1-9(14)11-3-2-10(8-12(11)15)13-4-6-16-7-5-13/h2-3,8,15H,4-7H2,1H3
InChI key
YHKSBKQXCWHTQL-UHFFFAOYSA-N
General description
Biochem/physiol Actions
Packaging
Warning
Other Notes
Willmore, E., et al. 2004. Blood,103, 4659.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service