Skip to Content
Merck
All Photos(1)

Key Documents

ENA424045637

Enamine

1-[(3-cyanophenyl)methyl]-5-oxopyrrolidine-2-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Molecular Weight:
244.25
MDL number:
Pricing and availability is not currently available.

Iupac Name

1-(3-cyanobenzyl)-5-oxoproline

Inchi Code

1S/C13H12N2O3/c14-7-9-2-1-3-10(6-9)8-15-11(13(17)18)4-5-12(15)16/h1-3,6,11H,4-5,8H2,(H,17,18)

InChI key

ZLNDUOFDNMCDSU-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-175150

MSDS

https://www.enaminestore.com/catalog/EN300-175150


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library