Skip to Content
Merck
All Photos(1)

Key Documents

900573

Sigma-Aldrich

(R)-2-Phenylpiperidine

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15N
CAS Number:
Molecular Weight:
161.24
UNSPSC Code:
12352200

Assay

95%

form

liquid

storage temp.

2-8°C

InChI

1S/C11H15N/c1-2-6-10(7-3-1)11-8-4-5-9-12-11/h1-3,6-7,11-12H,4-5,8-9H2/t11-/m1/s1

InChI key

WGIAUTGOUJDVEI-LLVKDONJSA-N

Application

Chiral building block developed using Liverpool ChiroChem-patented technology.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

219.2 °F

Flash Point(C)

104 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service