Skip to Content
Merck
All Photos(1)

Key Documents

766429

Sigma-Aldrich

(1R,4R,5R)-4,7,7-Trimethyl-6-thiabicyclo[3.2.1]octane

Synonym(s):

exo-(-)-4,7,7-Trimethyl-6-thiabicyclo[3.2.1]octane, Isothiocineole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18S
CAS Number:
Molecular Weight:
170.31
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

storage temp.

2-8°C

SMILES string

C[C@H]1CC[C@@H]2C[C@H]1SC2(C)C

InChI

1S/C10H18S/c1-7-4-5-8-6-9(7)11-10(8,2)3/h7-9H,4-6H2,1-3H3/t7-,8+,9+/m0/s1

InChI key

FAXNZPOZWCWYBD-DJLDLDEBSA-N

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

190.0 °F

Flash Point(C)

87.8 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service