Improving the Chromatographic Separation of DMB-Labeled Sialic Acids for the Comparison of Biosimilars to Reference Materials
Xiaoning Lu, Isil Yasa, Ben Cutak, Kevin Ray, David S. Bell
Reporter US Volume 33.2
Read more about
Abstract
Sialic acids are N- or O-substituted derivatives of neuraminic acid. They are important because they affect bioavailability, function, stability, and metabolism of glycoproteins. Two forms of sialic acid are commonly present in therapeutic glycoproteins: N-acetylneuraminic acid (NANA) and N-glycolylneuraminic acid (NGNA). One of the most common quantification methods involves releasing the sialic acids from the glycoprotein, derivatizing NANA and NGNA with 1,2-diamino-4, 5-methylenedioxybenzene dihydrochloride (DMB), followed by analysis by Reversed-Phase HPLC with fluorescence detection. This procedure is subject to interference from peaks originating from excess reagent and other derivatized impurities, limiting sensitivity and reproducibility. The objectives of this study were to develop a significantly improved HPLC-fluorescence method for DMB-NANA and DMB-NGNA, and to apply this method to compare two candidate biosimilar therapeutic proteins to their respective reference materials.
Experimental
LC optimization was performed using standard mixtures of DMB-NANA and DMB-NGNA using HPLC with fluorescence detection. Four column types of the same dimensions were evaluated: C18, F5, RP-Amide (all reversed-phase columns), and bare silica (HILIC column). Flow rate and column temperature were held constant at 0.2 mL/min and 30 °C, respectively. Mobile phase composition varied, but A and B solvents were always 0.1% formic acid in water and 0.1% formic acid in acetonitrile, respectively.
Sialic acids were released from glycoproteins, derivatized with DMB (Figure 1), and then analyzed at the optimal chromatographic conditions with fluorescence detection using excitation and emission wavelengths of 373 nm and 448 nm, respectively. Data were fit to external calibration curves. Results were expressed as the ratio of (moles of sialic acid) per (moles of protein).
![Derivatization of Sialic Acids with 1,2-Diamino-4,5-methylenedioxybenzene (DMB). A chemical process diagram illustrating the derivatization of sialic acids with 1,2-Diamino-4,5-methylenedioxybenzene (DMB). On the left side, a molecule labeled as “Sialylated glycoconjugate” undergoes hydrolysis by an enzyme called “NANase” to produce “Free sialic acid”. This free sialic acid is then subjected to DMB derivatization, resulting in a molecule labeled as “Labeled sialic acid” on the right side. Below this process flow, the chemical structure of 1,2-Diamino-4,5-methylenedioxybenzene (DMB) is depicted. The image is relevant for demonstrating the chemical modification of sialic acids which can be important in biochemical analysis and research.](/deepweb/assets/sigmaaldrich/marketing/global/images/technical-documents/articles/analytical-chemistry/small-molecule-hplc/derivatization-of-sialic/derivatization-of-sialic.jpg)
Figure 1. Derivatization of Sialic Acids with 1,2-Diamino-4,5-methylenedioxybenzene (DMB).
Results and Discussion
Figure 2 shows a representative chromatogram of the HPLC separation of the two common sialic acids, DMB-NANA and DMB-NGNA, on a C18 reversed-phase column. The two sialic acids are not completely baseline resolved, while the separation also suffers from a relatively long (25 min) run time and a ternary mobile phase composition. Furthermore, a later study showed that the sialic acids tend to co-elute with several derivatized interferences.
![Representative Chromatogram of HPLC Separation of DMB-Labeled Sialic Acids with an Ascentis<sup>®</sup> C18 Column CONDITIONS: column: Ascentis C18, 15 cm × 2.1 mm I.D., 3 µm (Product No. 581302-U); mobile phase: water:acetonitrile:methanol (84:9:7); gradient: isocratic, 25 minute run time. A representative chromatogram of High-Performance Liquid Chromatography (HPLC) separation of DMB-labeled sialic acids. The graph shows ‘Fluorescence’ on the y-axis and ‘Time (min)’ on the x-axis. There are four distinct peaks labeled ‘NGNA,’ ‘NANA,’ ‘Standard injection,’ and ‘Excess Reagents.’ The peaks for NGNA and NANA appear early, followed by a larger peak for Standard injection, and a very large peak for Excess Reagents towards the end. The chromatogram was obtained using an Ascentis® C18 column with specific conditions: 15 cm × 2.1 mm I.D., 3 µm (Product No. 581302-U); mobile phase: water:acetonitrile:methanol (84:9:7); gradient: isocratic, 25 minute run time.](/deepweb/assets/sigmaaldrich/marketing/global/images/technical-documents/articles/analytical-chemistry/small-molecule-hplc/chromatogram-of-hplc/chromatogram-of-hplc.jpg)
Figure 2. Representative Chromatogram of HPLC Separation of DMB-Labeled Sialic Acids with an Ascentis® C18 Column CONDITIONS: column: Ascentis C18, 15 cm × 2.1 mm I.D., 3 µm (Product No. 581302-U); mobile phase: water:acetonitrile:methanol (84:9:7); gradient: isocratic, 25 minute run time.
We screened several column chemistries (Table 1) in an attempt to develop an alternative method that would provide a shorter analysis time, better overall selectivity and a simpler binary mobile phase composition.
The column screening results in Figure 3 show that the RP-Amide column provides better selectivity for detecting the DMB-labeled sialic acids from the interferences. Therefore this column was chosen for further method development and optimization.
![HPLC separations of DMB-labeled sialic acid on different columns. Four chromatograms representing High-Performance Liquid Chromatography (HPLC) separations of DMB-labeled sialic acid on different columns. Each chromatogram is labeled with the column type: C18, F5, RP-Amide, and HILIC (Si). The x-axis represents time in minutes (0 to 10), and the y-axis represents detector response in arbitrary units. Each chromatogram shows two main peaks labeled ‘1. DMB-NGNA’ and ‘2. DMB-NANA,’ indicating the separation of these compounds. The C18 column shows peaks at approximately 1 and 2 minutes, the F5 column shows a large peak at around 4 minutes and a smaller peak just before it, the RP-Amide column displays sharp peaks at about 4 and 5 minutes, and the HILIC (Si) column presents closely eluting peaks around the same retention times as RP-Amide.](/deepweb/assets/sigmaaldrich/marketing/global/images/technical-documents/articles/analytical-chemistry/small-molecule-hplc/separations-of-dmb-labled/separations-of-dmb-labled.jpg)
Figure 3. HPLC separations of DMB-labeled sialic acid on different columns.
As can be seen in Figure 4, further optimization of the separation with the Ascentis Express RP-Amide column enabled baseline resolution of the two sialic acids, DMB-NGNA and DMB-NANA, with a 15-minute run time using a simple mobile phase composition (water and acetonitrile, each containing 0.1% formic acid). Importantly, the interferences are well resolved from the sialic acid analytes.
Besides the advantages of a shorter run time, better selectivity, and a simpler mobile phase composition, the newly developed method on the Ascentis Express RP-Amide column also provides a lower limit of quantification and a wider linear range than the original Ascentis C18 method (Table 2.)
![Optimized HPLC Separation of DMB-Labeled Sialic Acids with the RP-Amide Column CONDITIONS: column: Ascentis Express RP-Amide, 10 cm × 2.1 mm I.D., 2.7 µm (<a href="/product/supelco/53913U">53913-U</a>); mobile phase: [A] water; [B] acetonitrile, both with 0.1% formic acid; gradient: 0-1 min 6% B; 1.01-4 min 20%B; 4.01-12 min 6% B, total run time 15 min. Chromatogram showing optimized HPLC separation of DMB-labeled sialic acids with the RP-Amide Column. Two prominent peaks are labeled NGNA and NANA, indicating the presence of these sialic acids. There are also smaller peaks labeled as ‘Interferences’ and one at the beginning marked ‘Interference’. The x-axis is labeled ‘Time (min)’ ranging from 0 to 14 minutes, and the y-axis represents the detector response without specified units.](/deepweb/assets/sigmaaldrich/marketing/global/images/technical-documents/articles/analytical-chemistry/small-molecule-hplc/optimized-hplc/optimized-hplc.jpg)
Figure 4. Optimized HPLC Separation of DMB-Labeled Sialic Acids with the RP-Amide Column CONDITIONS: column: Ascentis Express RP-Amide, 10 cm × 2.1 mm I.D., 2.7 µm (53913-U); mobile phase: [A] water; [B] acetonitrile, both with 0.1% formic acid; gradient: 0-1 min 6% B; 1.01-4 min 20%B; 4.01-12 min 6% B, total run time 15 min.
Conc. (µg/mL) | RP-Amide Method % Accuracy NGNA | % Accuracy NANA | C18 Method % Accuracy NGNA | % Accuracy NANA |
---|---|---|---|---|
10 | 94% | 92% | 89% | 93% |
5 | 100% | 100% | 100% | 101% |
2.5 | 99% | 98% | 100% | 98% |
1.25 | 98% | 99% | 96% | 97% |
0.625 | 101% | 100% | 104% | 107% |
0.5 | 102% | 102% | 100% | 103% |
0.313 | 103% | 102% | 103% | 101% |
0.156 | 104% | 108% | 128% | 116% |
0.078 | 109% | 110% | 186% | 179% |
0.039 | 165% | 206% | 126% | 125% |
The newly developed RP-Amide method was validated by determining the sialic acid content in erythropoietin (EPO), a glycoprotein. Figure 5 shows the successful detection of NANA at different quantities (2.5, 5.0 and 10 µg) of EPO with minimal interference. The determined molar amounts of NANA per mole EPO are consistent between the three levels of EPO, all passing the specification (>10 mol NANA/mol EPO).
![Determination of the sialic acids of EPO. Graphical representation of the determination of sialic acids in Erythropoietin (EPO) samples. The image shows a chromatogram with a y-axis labeled ‘Fluorescence’ and an x-axis labeled ‘Time (min).’ Three peaks are visible, corresponding to different amounts of EPO: 2.4 µg, 5.0 µg, and 10 µg, with the highest peak at the 10 µg mark. Below the graph is a table titled ‘Sample’ with three rows for EPO-2.4, EPO-5, and EPO-10. Columns include ‘EPO Amount (µg),’ ‘mol NANA / mol EPO,’ and ‘Meet Specification,’ all entries in the last column read ‘Yes.’](/deepweb/assets/sigmaaldrich/marketing/global/images/technical-documents/articles/analytical-chemistry/small-molecule-hplc/determination-of-sialic-acids/determination-of-sialic-acids.jpg)
Figure 5. Determination of the sialic acids of EPO.
The RP-Amide method has been successfully applied to determine and compare sialic acid levels in biosimilar candidates to their authentic references. Figure 6 shows that the NANA level in TNFr-FC (Enbrel®) is about 30% lower than in the biosimilar candidate compared to its authentic reference. While Figure 7 shows that the NGNA content in the Erbitux® biosimilar candidate is almost twice that of its authentic reference.
![Quantitative comparison of the NANA content in TNFr-FC (Enbrel<sup>®</sup>) biosimilar candidate to its authentic reference. Graphical representation of the NANA content in TNFr-FC (Enbrel®) biosimilar candidate compared to its authentic reference. The graph displays two peaks, with the ‘Authentic Reference’ peak being significantly higher than the ‘Biosimilar Candidate’ peak, indicating a higher NANA content. Below the graph, a table shows the determined NANA in TNFr-FC with ‘Authentic Reference’ having 15.8 mol NANA/mol protein and ‘Biosimilar Candidate’ having 10.3 mol NANA/mol protein.](/deepweb/assets/sigmaaldrich/marketing/global/images/technical-documents/articles/analytical-chemistry/small-molecule-hplc/quantitative-comparison/quantitative-comparison.jpg)
Figure 6. Quantitative comparison of the NANA content in TNFr-FC (Enbrel®) biosimilar candidate to its authentic reference.
![Quantitative comparison of NGNA content in erbitux biosimilar candidate to its authentic reference. Graphical representation of NGNA content comparison between Erbitux and its biosimilar candidate. The graph displays two overlaid chromatograms with peaks indicating NGNA levels. The ‘Erbitux’ chromatogram shows higher peaks compared to the ‘Biosimilar Candidate,’ suggesting a quantitative difference in NGNA content. Below the graph, a table titled ‘Determined NGNA in Erbitux’ lists the sample, protein amount, and mol NGNA/mol protein ratios for both Erbitux and the biosimilar candidate.](/deepweb/assets/sigmaaldrich/marketing/global/images/technical-documents/articles/analytical-chemistry/small-molecule-hplc/quanntitative-comparasion-of-ngna/quanntitative-comparasion-of-ngna.jpg)
Figure 7. Quantitative comparison of NGNA content in erbitux biosimilar candidate to its authentic reference.
Conclusions
A new HPLC method employing an Ascentis Express RP-Amide column has been developed to determine DMB-labeled NGNA and NANA sialic acids released from biotherapeutic proteins. Compared to the original method using a C18 column, the new method offers the following advantages:
- Simpler mobile phase: water and acetonitrile containing 0.1% formic acid
- Better resolution for DMB-NGNA and DMB-NANA sialic acids
- Shorter run time
- Improved peak shapes
- Wider linear range for quantification
- Lower limit of quantification
Trademarks
Ascentis is a registered trademark of Sigma-Aldrich Co. LLC
Enbrel is a registered trademark of Immunex Corp.
Erbutix is a registered trademark of ImClone LLC