Skip to Content
Merck
All Photos(3)

Key Documents

V900394

Sigma-Aldrich

5-Fluorouracil

Vetec, reagent grade, ≥99%

Synonym(s):

2,4-Dihydroxy-5-fluoropyrimidine, 5-FU, 5-Fluoro-2,4(1H,3H)-pyrimidinedione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H3FN2O2
CAS Number:
Molecular Weight:
130.08
Beilstein:
127172
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

Assay

≥99%

form

powder

mp

282-286 °C (dec.) (lit.)

solubility

1 M NH4OH: 50 mg/mL, clear, colorless to faintly yellow

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

FC1=CNC(=O)NC1=O

InChI

1S/C4H3FN2O2/c5-2-1-6-4(9)7-3(2)8/h1H,(H2,6,7,8,9)

InChI key

GHASVSINZRGABV-UHFFFAOYSA-N

Gene Information

human ... TYMS(7298)

Looking for similar products? Visit Product Comparison Guide

Application

5-Fluorouracil (5-FU) can be used as a precursor for the preparation of 5-FU derivatives such as 1-acyloxyalkyl-5-fluorouracil, 3-acyloxyalkyl-5-fluorouracil, 1, 3-bis (acyloxyalkyl)-5-fluorouracil, and 1-hexylcarbamoyl-5-fluorouracil (HCFU), as potent antitumor agents.[1][2] It can be also used in the synthesis of 5-5FU/diazeniumdiolate conjugate, which exhibits a greater cytotoxic effect than fluorouracil on cancer cells.[3]

Biochem/physiol Actions

5-Fluorouracil (5-FU) is a potent antitumor agent that affects pyrimidine synthesis by inhibiting thymidylate synthetase, thus depleting intracellular dTTP pools. It is metabolized to ribonucleotides and deoxyribonucleotides, which can be incorporated into RNA and DNA.[4][5] Treatment of cells with 5-FU leads to an accumulation of cells in S-phase and has been shown to induce p53 dependent apoptosis.[6][7]

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

5-Fluorouracil derivatives. I. The synthesis of 1-carbamoyl-5-fluorouracils
Ozaki S, et al.
Bulletin of the Chemical Society of Japan, 50(9), 2406-2412 (1977)
Handbook of Reagents for Organic Synthesis, Sulfur-Containing Reagents (2004)
Synthesis and cytotoxicity of 5-fluorouracil/diazeniumdiolate conjugates
Cai TB, et al.
Bioorganic & Medicinal Chemistry, 11(23), 4971-4975 (2003)
Molecular Mechanisms Of Drug Action (1994)
5-fluorouracil activation of p53 involves an MDM2-ribosomal protein interaction.
Sun X X, et al.
The Journal of Biological Chemistry, 282(11), 8052-8059 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service