Skip to Content
Merck
All Photos(1)

Key Documents

V800170

Sigma-Aldrich

Ethylenediaminetetraacetic acid disodium salt dihydrate

AR, ≥99%

Synonym(s):

Disodium ethylenediaminetetraacetate dihydrate, EDTA disodium salt, EDTA-Na2, Edathamil, Edetate disodium salt dihydrate, Sequestrene Na2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N2Na2O8 · 2H2O
CAS Number:
Molecular Weight:
372.24
Beilstein:
3900609
EC Number:
UNSPSC Code:
12352107
Pricing and availability is not currently available.

grade

AR

product line

Vetec

Assay

≥99%

mp

248 °C (dec.) (lit.)

solubility

H2O: 100 g/L at 20 °C

SMILES string

O.O.O.O.[Na+].[Na+].[Na+].[Na+].OC(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC(O)=O.OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

InChI

1S/C10H16N2O8.2Na.2H2O/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;;2*1H2/q;2*+1;;/p-2

InChI key

OVBJJZOQPCKUOR-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Chelator of divalent cations. Inhibits enzymes, such as metalloproteases, that require divalent cations for activity.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - STOT RE 2 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suli Huang et al.
PloS one, 10(2), e0117007-e0117007 (2015-02-07)
microRNA (miRNA) plays a role in the pathogenesis of ischemic stroke, and single nucleotide polymorphisms in miRNA genes may contribute to disease susceptibility. However, the effect of miR-146a, miR-196a2, and miR-499 polymorphisms on ischemic stroke susceptibility has been rarely reported.
Lisa M Ryno et al.
Biomacromolecules, 15(8), 2944-2951 (2014-07-18)
End-functionalized macromolecular starch reagents, prepared by reductive amination, were grafted onto a urethane-linked polyester-based backbone using copper-catalyzed azide-alkyne cycloaddition (CuAAC) chemistry to produce novel amphiphilic hybrid graft copolymers. These copolymers represent the first examples of materials where the pendant chains
Haiyan Zhang et al.
Analytica chimica acta, 770, 140-146 (2013-03-19)
A novel method for selective determination of Cr(III) and Cr(VI) in environmental water samples was developed based on target-induced fluorescence quenching of glutathione-stabilized gold nanoclusters (GSH-Au NCs). Fluorescent GSH-Au NCs were synthesized by a one-step approach employing GSH as reducing/protecting
Gervasio A Lamas et al.
JAMA, 309(12), 1241-1250 (2013-03-28)
Chelation therapy with disodium EDTA has been used for more than 50 years to treat atherosclerosis without proof of efficacy. To determine if an EDTA-based chelation regimen reduces cardiovascular events. Double-blind, placebo-controlled, 2 × 2 factorial randomized trial enrolling 1708
Seung Hyeok Seok et al.
Journal of applied toxicology : JAT, 33(10), 1089-1096 (2013-02-15)
Zinc oxide (ZnO) nanoparticles (NPs) are used in diverse applications ranging from paints and cosmetics to biomedicine and food. Although micron-sized ZnO is a traditional food supplement, ZnO NPs are an unknown public health risk because of their unique physicochemical

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service