Skip to Content
Merck
All Photos(1)

Key Documents

V800048

Sigma-Aldrich

Ammonium iron(II) sulfate hexahydrate

LR, ≥98.5%

Synonym(s):

Ammonium ferrous sulfate hexahydrate, Ammonium iron(II) sulfate, Mohr’s salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2Fe(SO4)2 · 6H2O
CAS Number:
Molecular Weight:
392.14
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
LR
form:
crystalline powder

grade

LR

product line

Vetec

Assay

≥98.5%

form

crystalline powder

mp

100 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤0.002%

cation traces

Fe3+: ≤0.03%
Pb: ≤0.003%

SMILES string

N.N.O.O.O.O.O.O.[Fe++].OS([O-])(=O)=O.OS([O-])(=O)=O

InChI

1S/Fe.2H3N.2H2O4S.6H2O/c;;;2*1-5(2,3)4;;;;;;/h;2*1H3;2*(H2,1,2,3,4);6*1H2/q+2;;;;;;;;;;/p-2

InChI key

MQLVWQSVRZVNIP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mayara Marinovic Baccan et al.
Journal of inorganic biochemistry, 107(1), 34-39 (2011-12-17)
The antioxidant activity of flavonoids may involve their ability to complex body iron in non-redox-active forms. In this study, it was found that the catechol flavonoids rutin and quercetin are able to suppress redox-active labile plasma iron (LPI) in both
Seong-Ryong Lee et al.
Phytotherapy research : PTR, 17(3), 206-209 (2003-04-03)
The aim of this study was to compare the protective effects of green tea polyphenol (-)-epigallocatechin gallate (EGCG) and other well-known antioxidants on the lipid peroxidation in gerbil brain homogenates. Oxidative stress was induced by H2O2 (10 mM) or ferrous
Hyung Lee et al.
Journal of neurosurgical anesthesiology, 17(3), 144-148 (2005-07-23)
This study compared the effectiveness of propofol with that of trolox and melatonin for reduction of lipid peroxidation in vitro. Lipid peroxidation was induced by addition of kainic acid (KA; 10 mM), hydrogen peroxide (H2O2; 10 mM), or ferrous ammonium
Cinzia Signorini et al.
Pediatric research, 63(3), 287-291 (2008-02-22)
Nonprotein-bound iron (NPBI) and F2-isoprostanes, reliable markers of oxidative stress, are increased in plasma of newborns and inversely correlated to the gestational age. Because NPBI represents a pro-oxidant stimulus in plasma, we test the hypothesis that the entity of lipid
M K Semwal et al.
Physics in medicine and biology, 47(12), N179-N183 (2002-07-18)
The FBX aqueous chemical dosimeter contains 0.2 mol m(-3) ferrous ammonium sulphate, 5.0 mol m(-3) benzoic acid and 0.20 mol m(-3) xylenol orange in 40.0 mol m(-3) sulphuric acid. The dosimeter can measure photon and electron doses in the range

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service