Skip to Content
Merck
All Photos(1)

Key Documents

V800047

Sigma-Aldrich

Ammonium iron(II) sulfate hexahydrate

AR, ≥99%

Synonym(s):

Ammonium ferrous sulfate hexahydrate, Ammonium iron(II) sulfate, Mohr’s salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2Fe(SO4)2 · 6H2O
CAS Number:
Molecular Weight:
392.14
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
AR
form:
crystalline powder
Pricing and availability is not currently available.

grade

AR

product line

Vetec

Assay

≥99%

form

crystalline powder

pH

3-5

mp

100 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤0.001%

cation traces

Cu: ≤0.002%
Fe3+: ≤0.02%
K: ≤0.02%
Mn: ≤0.03%
Na: ≤0.02%
Zn: ≤0.005%

SMILES string

N.N.O.O.O.O.O.O.[Fe++].OS([O-])(=O)=O.OS([O-])(=O)=O

InChI

1S/Fe.2H3N.2H2O4S.6H2O/c;;;2*1-5(2,3)4;;;;;;/h;2*1H3;2*(H2,1,2,3,4);6*1H2/q+2;;;;;;;;;;/p-2

InChI key

MQLVWQSVRZVNIP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ashish Pathak et al.
Journal of hazardous materials, 171(1-3), 273-278 (2009-07-10)
The potential of indigenous iron-oxidizing microorganisms enriched at initial neutral pH of the sewage sludge for bioleaching of heavy metals was investigated at initial neutral pH of the sludge using ammonium ferrous sulfate (FAS) and ferrous sulfate (FS) as an
Cinzia Signorini et al.
Pediatric research, 63(3), 287-291 (2008-02-22)
Nonprotein-bound iron (NPBI) and F2-isoprostanes, reliable markers of oxidative stress, are increased in plasma of newborns and inversely correlated to the gestational age. Because NPBI represents a pro-oxidant stimulus in plasma, we test the hypothesis that the entity of lipid
Mayara Marinovic Baccan et al.
Journal of inorganic biochemistry, 107(1), 34-39 (2011-12-17)
The antioxidant activity of flavonoids may involve their ability to complex body iron in non-redox-active forms. In this study, it was found that the catechol flavonoids rutin and quercetin are able to suppress redox-active labile plasma iron (LPI) in both
Hyung Lee et al.
Journal of neurosurgical anesthesiology, 17(3), 144-148 (2005-07-23)
This study compared the effectiveness of propofol with that of trolox and melatonin for reduction of lipid peroxidation in vitro. Lipid peroxidation was induced by addition of kainic acid (KA; 10 mM), hydrogen peroxide (H2O2; 10 mM), or ferrous ammonium
M K Semwal et al.
Physics in medicine and biology, 47(12), N179-N183 (2002-07-18)
The FBX aqueous chemical dosimeter contains 0.2 mol m(-3) ferrous ammonium sulphate, 5.0 mol m(-3) benzoic acid and 0.20 mol m(-3) xylenol orange in 40.0 mol m(-3) sulphuric acid. The dosimeter can measure photon and electron doses in the range

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service