524633
Phosphatase Inhibitor Cocktail Set IV, Lyophilized
The Phosphatase Inhibitor Cocktail Set IV, Lyophilized controls the activity of Phosphatase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352200
form:
lyophilized solid
storage temp.:
2-8°C
Recommended Products
Quality Level
form
lyophilized solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
desiccated (hygroscopic)
protect from light
shipped in
ambient
storage temp.
2-8°C
General description
A cocktail of three phosphatase inhibitors for the inhibition of both serine/threonineand alkaline phosphatases. Each vial, when reconstituted with 1 ml DMSO contains 500 µM Cantharidin (Cat. No. 210155), 2.5 mM (-)-p-Bromotetramisole oxalate, and 1 µ Calyculin A, Discodermia calyx (Cat. No. 208851).
Packaging
Packaged under inert gas
Warning
Toxicity: Highly Toxic (H)
Reconstitution
Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.
Reconstitute with 1 ml of 95% DMSO. Slight warming may be required for complete dissolution; warm only if needed.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 2 Oral - Acute Tox. 4 Dermal
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Monica M Sauter et al.
Experimental eye research, 204, 108436-108436 (2021-01-14)
The goal of this study was to determine the expression and distribution of the host restriction factors (RFs) TRIM5α and TRIM11 in non-human primate (NHP) neural retina tissue and the human Muller cell line MIO-M1. In addition, experiments were performed
Ninan Zhang et al.
ACS omega, 9(21), 22754-22763 (2024-06-03)
Curcumin, a compound derived from turmeric, is traditionally utilized in East Asian medicine for treating various health conditions, including epilepsy. Despite its involvement in numerous cellular signaling pathways, the specific mechanisms and targets of curcumin in epilepsy treatment have remained
Tong Wang et al.
BMC genomics, 22(1), 840-840 (2021-11-21)
Sanghuangporus sanghuang is a well-known traditional medicinal mushroom associated with mulberry. Despite the properties of this mushroom being known for many years, the regulatory mechanisms of bioactive compound biosynthesis in this medicinal mushroom are still unclear. Lysine malonylation is a
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service