Skip to Content
Merck
All Photos(2)

Key Documents

426032

Sigma-Aldrich

Boc-3-iodo-D-Ala-OMe

99%

Synonym(s):

N-(tert-Butoxycarbonyl)-3-iodo-D-alanine methyl ester, Boc-β-iodo-D-Ala-OMe, Boc-3-iodo-D-alanine methyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
RM 1,258.00

RM 1,258.00


Estimated to ship on11 April 2025


Request a Bulk Order

Select a Size

Change View
5 G
RM 1,258.00

About This Item

Linear Formula:
ICH2CH[NHCO2C(CH3)3]CO2CH3
CAS Number:
Molecular Weight:
329.13
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

RM 1,258.00


Estimated to ship on11 April 2025


Request a Bulk Order

Quality Level

Assay

99%

form

solid

reaction suitability

reaction type: Boc solid-phase peptide synthesis

mp

55-59 °C (lit.)

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

COC(=O)[C@@H](CI)NC(=O)OC(C)(C)C

InChI

1S/C9H16INO4/c1-9(2,3)15-8(13)11-6(5-10)7(12)14-4/h6H,5H2,1-4H3,(H,11,13)/t6-/m1/s1

InChI key

UGZBFCCHLUWCQI-ZCFIWIBFSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service