Merck
Prostaglandin E2 promotes features of replicative senescence in chronically activated human CD8+ T cells.
PloS one (2014-06-12)
Jennifer P Chou, Christina M Ramirez, Danielle M Ryba, Megha P Koduri, Rita B Effros
RESUMEN
Prostaglandin E2 (PGE2), a pleiotropic immunomodulatory molecule, and its free radical catalyzed isoform, iso-PGE2, are frequently elevated in the context of cancer and chronic infection. Previous studies have documented the effects of PGE2 on the various CD4+ T cell functions, but little is known about its impact on cytotoxic CD8+ T lymphocytes, the immune cells responsible for eliminating virally infected and tumor cells. Here we provide the first demonstration of the dramatic effects of PGE2 on the progression of human CD8+ T cells toward replicative senescence, a terminal dysfunctional state associated multiple pathologies during aging and chronic HIV-1 infection. Our data show that exposure of chronically activated CD8+ T cells to physiological levels of PGE2 and iso-PGE2 promotes accelerated acquisition of markers of senescence, including loss of CD28 expression, increased expression of p16 cell cycle inhibitor, reduced telomerase activity, telomere shortening and diminished production of key cytotoxic and survival cytokines. Moreover, the CD8+ T cells also produced higher levels of reactive oxygen species, suggesting that the resultant oxidative stress may have further enhanced telomere loss. Interestingly, we observed that even chronic activation per se resulted in increased CD8+ T cell production of PGE2, mediated by higher COX-2 activity, thus inducing a negative feedback loop that further inhibits effector function. Collectively, our data suggest that the elevated levels of PGE2 and iso-PGE2, seen in various cancers and HIV-1 infection, may accelerate progression of CD8+ T cells towards replicative senescence in vivo. Inhibition of COX-2 activity may, therefore, provide a strategy to counteract this effect.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Dimetilsulfóxido, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimetilsulfóxido, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimetilsulfóxido, for molecular biology
Sigma-Aldrich
Dimetilsulfóxido, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimetilsulfóxido, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimetilsulfóxido, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimetilsulfóxido, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimetilsulfóxido, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Forskolina, from Coleus forskohlii, ≥98% (HPLC), powder
Sigma-Aldrich
Forskolina, For use in molecular biology applications
Sigma-Aldrich
Dimetilsulfóxido, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Prostaglandin E2, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Dimetilsulfóxido, PCR Reagent
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate, ≥98.5% (HPLC), powder
Sigma-Aldrich
Dimetilsulfóxido, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimetilsulfóxido, anhydrous, ≥99.9%
Sigma-Aldrich
Prostaglandin E2, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Prostaglandin E2, ≥93% (HPLC), synthetic
USP
Dimetilsulfóxido, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate tris salt, ≥97% (HPLC), powder
Sigma-Aldrich
Dimetilsulfóxido, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Supelco
Dimetilsulfóxido, analytical standard
Sigma-Aldrich
Dimetilsulfóxido, Vetec, reagent grade, 99%
Supelco
Forskolina, analytical standard
Sigma-Aldrich
AH6809, ≥98%, crystalline solid or supercooled liquid
Supelco
Dimetilsulfóxido, for inorganic trace analysis, ≥99.99995% (metals basis)
Dimetilsulfóxido, European Pharmacopoeia (EP) Reference Standard
Dinoprostone, European Pharmacopoeia (EP) Reference Standard