Merck
  • SKLB316, a novel small-molecule inhibitor of cell-cycle progression, induces G2/M phase arrest and apoptosis in vitro and inhibits tumor growth in vivo.
SKLB316, a novel small-molecule inhibitor of cell-cycle progression, induces G2/M phase arrest and apoptosis in vitro and inhibits tumor growth in vivo.
Cancer letters (2014-10-11)
Yong Xia, Qian Lei, Yongxia Zhu, Tinghong Ye, Ningyu Wang, Guobo Li, Xuanhong Shi, Yantong Liu, Bin Shao, Tao Yin, Lifeng Zhao, Wenshuang Wu, Xuejiao Song, Ying Xiong, Yuquan Wei, Luoting Yu
RESUMEN
Benzothiazole derivatives have received considerable attentions for their potencies in cancer therapy. In the present study, we reported that SKLB316, a novel synthesized benzothiazole derivative, exhibits activities to inhibit colorectal and pancreatic cancer in vitro and in vivo by inducing G2/M cell cycle arrest and apoptosis. In vitro, it exhibited significant anti-proliferative activities against human cancer cells derived from different histotypes including the colorectal cancer cell line HCT116 and pancreatic cancer cell line CFPAC-1. We chose these cell lines to study the possible anti-tumor mechanism because they are sensitive to SKLB316 treatment. Flow cytometry assays showed that SKLB316 could induce G2/M cell cycle arrest. Mechanistically, SKLB316 could decrease the activities of cdc2/cyclin B1 complex, including decreasing the synthesis of cyclin B1, cdc2 and cdc25c, while accumulating the levels of phosphorylated cdc2 (Tyr15) and checkpoint kinase 2. SKLB316 could also decrease the level of cyclin E and A2. Moreover, SKLB316 could induce cancer cell apoptosis, which was associated with activation of caspase 9, downregulation of Bcl-2 and upregulation of Bax. SKLB316 could also decrease the mitochondrial membrane potential and induce the generation of reactive oxygen species in cells. The results implied that SKLB316 may induce apoptosis via the mitochondria-mediated apoptotic pathway. Moreover, SKLB316 could suppress the growth of established colorectal and pancreatic cancer tumors in nude mice without causing obvious side effects. TUNEL assays confirmed that SKLB316 could also induce tumor cell apoptosis in vivo. Taken together, these findings demonstrate the potential value of SKLB316 as a novel anti-tumor drug candidate.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Alcohol etílico puro, for molecular biology
Sigma-Aldrich
Dimetilsulfóxido, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimetilsulfóxido, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimetilsulfóxido, for molecular biology
Sigma-Aldrich
Alcohol etílico puro, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Dimetilsulfóxido, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimetilsulfóxido, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Alcohol etílico puro, 200 proof
Sigma-Aldrich
Dimetilsulfóxido, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Alcohol etílico puro, 200 proof, meets USP testing specifications
Sigma-Aldrich
Dimetilsulfóxido, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Alcohol etílico puro, for molecular biology
Sigma-Aldrich
Dimetilsulfóxido, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Yoduro de propidio, ≥94.0% (HPLC)
Sigma-Aldrich
Fluoruro de fenilmetansulfonilo, ≥98.5% (GC)
Sigma-Aldrich
Etanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Dimetilsulfóxido, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Fluoruro de fenilmetansulfonilo, ≥99.0% (T)
Sigma-Aldrich
Dimetilsulfóxido, PCR Reagent
Sigma-Aldrich
Alcohol etílico puro, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Dimetilsulfóxido, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Etanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Alcohol etílico puro, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Dimetilsulfóxido, anhydrous, ≥99.9%
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Ethanol solution, 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Etanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Etanol, BioUltra, for molecular biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
USP
Dimetilsulfóxido, United States Pharmacopeia (USP) Reference Standard
USP
Etanol, United States Pharmacopeia (USP) Reference Standard