Skip to Content
Merck
All Photos(1)

Key Documents

R8401

Sigma-Aldrich

Rose Bengal diacetate

Synonym(s):

Spiro[isobenzofuran-1(3H), 9′-[9H]xanthen]-3-one, 3′,6′-bis(Acetyloxy)-4,5,6, 7-tetrachloro-2′,4′,5′,7′-spiro[isobenzofuran-1(3H), 9′-[9H]xanthen]-3-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H8Cl4I4O7
CAS Number:
Molecular Weight:
1057.75
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Assay

≥98% (TLC)

form

powder

mol wt

1057.75

εmax

8.0-10.0 at 312-313 nm in methanol

application(s)

diagnostic assay manufacturing
hematology
histology

shipped in

dry ice

storage temp.

−20°C

SMILES string

CC(OC1=C(I)C=C2C(OC(C(I)=C(OC(C)=O)C(I)=C3)=C3C24OC(C5=C4C(Cl)=C(Cl)C(Cl)=C5Cl)=O)=C1I)=O

InChI

1S/C24H12Cl4O7/c1-9(29)32-11-3-5-13-15(7-11)34-16-8-12(33-10(2)30)4-6-14(16)24(13)18-17(23(31)35-24)19(25)21(27)22(28)20(18)26/h3-8H,1-2H3

InChI key

HHXVUGCSLBZDTM-UHFFFAOYSA-N

General description

A cell-permeable generator of singlet oxygen.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Systematic Study of Parameters Influencing the Action of Rose Bengal with Visible Light on Bacterial Cells: Comparison Between the Biological Effect and Singlet-Oxygen Production
M. C. Schaefer
Schweizerische Monatsschrift fur Zahnheilkunde = Revue Mensuelle Suisse D'Odonto-stomatologie / SSO (2000)
Jimmy D. Bartlett
Clinical Ocular Pharmacology null
Comparison of two methods to identify live benthic foraminifera:
A test between Rose Bengal and CellTracker Green with
implications for stable isotope paleoreconstructions
Joan M. Bernhard
Palladium Reagents in Organic Synthesis (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service