Skip to Content
Merck
All Photos(2)

Key Documents

34338

Supelco

Drazoxolon

PESTANAL®, analytical standard

Synonym(s):

4-(2-Chlorophenylhydrazono)-3-methyl-5-isoxazolone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8ClN3O2
CAS Number:
Molecular Weight:
237.64
Beilstein:
1216595
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC1=NOC(=O)\C1=N/Nc2ccccc2Cl

InChI

1S/C10H8ClN3O2/c1-6-9(10(15)16-14-6)13-12-8-5-3-2-4-7(8)11/h2-5,12H,1H3/b13-9-

InChI key

OOTHTARUZHONSW-LCYFTJDESA-N

Looking for similar products? Visit Product Comparison Guide

General description

Drazoxolon is an oxazole fungicide[1] effective against seed and soil-borne diseases.[2]

Application

Drazoxolon may be used as an analytical reference standard for the determination of the analyte in fruit wine[3] and foodstuffs[4] by various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multi-residue detection of pesticides in juice and fruit wine: A review of extraction and detection methods.
Jin B, et al.
Food Research International, 46(1), 399-409 (2012)
Pesticide residue analysis in foodstuffs applying capillary gas chromatography with atomic emission detection State-of-the-art use of modified multimethod S19 of the Deutsche Forschungsgemeinschaft and automated large-volume injection with programmed-temperature vaporization and solvent venting.
Stan H, et al.
Journal of Chromatography A, 750(1-2), 369-390 (1996)
Comparison of stir bar sorptive extraction and membrane-assisted solvent extraction for the ultra-performance liquid chromatographic determination of oxazole fungicide residues in wines and juices.
Vi?as P, et al.
Journal of Chromatography A, 1194(2), 178-183 (2008)
Polarographic determination of some azomethine-containing pesticides.
Smyth MR and Osteryoung JG
Analytical Chemistry, 50(12), 1632-1637 (1978)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service