Skip to Content
Merck
All Photos(1)

Key Documents

HG0000C1

Millipore

Nutrient Agar

pH 7.1±0.2 (25 °C, after autoclaving)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116105

composition

Agar, 15.0 g/L
Meat Extract, 1.0 g/L
Peptone, 5.0 g/L
Sodium Chloride, 8.0 g/L
Yeast Extract, 2.0 g/L

packaging

pkg of 12 × 500 g (plastic bottles)

pH

7.1±0.2 (25 °C, after autoclaving)

storage temp.

15-25°C

Application

A general purpose media suitable for the cultivation of a wide range of non-fastidious organisms.

Preparation Note

Suspend 31g in 1 litre demineralised water. Boil whilst stirring until completely dissolved. Autoclave at 121°C for 15 minutes. Cool agar to 45 - 50°C, mix well and pour plates. If required, 5 - 10% blood may be added to the cooled, sterile agar before pouring the plates.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service