Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SLW1515/01D

Pyrex® Reagent bottles, round bottom with reusable screw caps

capacity 25 mL

Synonym(s):

Pyrex, SciLabware 1515/01D

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.17

material

PTFE seal
Pyrex bottle
black PBT cap
colorless bottle
round bottle
round bottom
white graduation

Joint

Joints threaded neck (SVL20)

feature

with cap

packaging

pack of 10 ea

manufacturer/tradename

SciLabware 1515/01D

bottle diam. × H

35 mm × 65 mm

cap size

20 mm

capacity

25 mL

Looking for similar products? Visit Product Comparison Guide

General description

  • Complies with ISO 4796-1
  • Ideal for storage of reagents, analytical standards and culture media etc.
  • Manufactured from Pyrex borosilicate glass
  • Excellent resistance to chemical attack and high temperatures
  • Supplied with PBT screwcaps having a PTFE faced sealing disc
  • Withstands hot air sterilisation up to 180 deg. C
  • Autoclavable
  • White graduations and marking spot

Legal Information

PYREX is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service