Key Documents
SLW1516/01RED
Pyrex® Media-Lab Bottles, with high temperature cap and pouring ring, with printed trace code
capacity 25 mL
Synonym(s):
Pyrex, SciLabware 1516/01RED
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
material
PTFE seal
red polyester cap (high temperature thermoplastic)
white graduation
Joint
Joints threaded neck (GL25)
feature
graduations
with cap
with pouring ring
with printed trace code
packaging
pack of 10 ea
manufacturer/tradename
SciLabware 1516/01RED
capacity
25 mL
diam. × H
36 mm × 70 mm
General description
  • Bottles complete with thermoplastic polyester cap, PTFE sealing disc and pouring ring
  • Complies with ISO 4796
  • Printed with Trace Code for downloadable batch certificate
  • Batch certificate available to download from www.scilabware.com
  • Withstands hot air sterilisation up to 180 deg. C
  • PTFE sealing disc resists chemical attack
  • White graduations and marking spot
  • Manufactured from temperature and chemically resistant borosilicate glass
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)