Key Documents
BMSBSH5002EUK
Sigma-Aldrich
myBlock ll digital dual chamber dry bath without blocks
AC/DC input 230 V AC, UK plug
About This Item
UNSPSC Code:
41102404
NACRES:
NB.76
manufacturer/tradename
Benchmark Scientific BSH5002-E-UK
parameter
5-105 °C temp. range
capacity
, holds 2 blocks
General description
Benchmark′s myBlock series is a "next generation" product line that introduces several major advantages over traditional dry baths. They are the first digital dry baths to include advanced microprocessor controls, timed or continuous operation and a removable hinged lid. In addition, an optional external temperature feedback probe can be positioned directly inside a sample tube, providing real-time control and monitoring of actual sample temperature.
  • Digital temperature control (set and walk away)
  • Hinged lid (improves uniformity & conserves energy)
  • External (optional) temperature probe (measures actual sample temperature)
  • Available with Quick-Flip universal blocks (supports all common tube sizes, PCR strips and PCR plates)
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)