Key Documents
Z742524
Block for digital dry bath and MyBlock
Holds 12 x 15-16 mm tubes
About This Item
UNSPSC Code:
41102404
NACRES:
NB.22
feature
Holds 12 x 15-16 mm tubes
packaging
pack of 1 ea
manufacturer/tradename
Benchmark Scientific BSW1516
General description
For use with Benchmark Digital Dry Bath
  • Material: high grade aluminum
  • Precision machined to match the tube shape and size, providing unsurpassed temperature uniformity
Legal Information
MyBlock is a trademark of Benchmark Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)