跳转至内容
Merck
  • Thymidine phosphorylase gene mutations in MNGIE, a human mitochondrial disorder.
Thymidine phosphorylase gene mutations in MNGIE, a human mitochondrial disorder.
Science (New York, N.Y.) (1999-01-29)
I Nishino, A Spinazzola, M Hirano
摘要
Mitochondrial neurogastrointestinal encephalomyopathy (MNGIE) is an autosomal recessive human disease associated with multiple deletions of skeletal muscle mitochondrial DNA (mtDNA), which have been ascribed to a defect in communication between the nuclear and mitochondrial genomes. Examination of 12 MNGIE probands revealed homozygous or compound-heterozygous mutations in the gene specifying thymidine phosphorylase (TP), located on chromosome 22q13.32-qter. TP activity in leukocytes from MNGIE patients was less than 5 percent of controls, indicating that loss-of-function mutations in TP cause the disease. The pathogenic mechanism may be related to aberrant thymidine metabolism, leading to impaired replication or maintenance of mtDNA, or both.
材料
货号
品牌
产品描述
Sigma-Aldrich
胸苷磷酸化酶,重组 来源于大肠杆菌, recombinant, expressed in E. coli, buffered aqueous solution, ≥500 units/mL
Sigma-Aldrich
胸苷磷酸化酶,重组 来源于大肠杆菌, recombinant, expressed in E. coli, buffered aqueous solution, ≥900 units/mL, 0.2 μm filtered