侧向层析滤膜
Hi-Flow™ Plus侧向层析膜是体外诊断检测试剂与试剂盒的理想选择,提供出色的结果一致性和检测性能。针对侧向层析检测专门优化,Hi-Flow™ Plus膜按照最严格的层析膜标准制造,超过市面上其他品牌,提供无与伦比的表面质量。特别设计的Hi-Flow™ Plus膜提供一致性结果、可信的批间性能、固定的检测时间、以及非特异性背景信号极低的明晰捕获/检测线。Hi-Flow™ Plus膜适用于各种侧向层析检测类型,并提供多种速度和规格的便利选择。
为侧向层析检测选择合适的原材料
侧向层析膜是侧向层析试纸条唯一最重要的材料。膜的理化性质影响毛细流动性。毛细流动性转而影响试剂沉积、检测灵敏度、检测特异性和检测线一致性。膜的其他物理性质影响与试纸条的装配。如需了解更多信息,可阅读我们的开发指南快速侧向层析试纸条——产品开发的考虑因素,深入解读侧向层析试纸条设计的关键要素,尤其是材料的选用及其与试剂的组装和生产工艺。了解这些设计原则有助于把握侧向层析检测的快速开发框架。
Hi-Flow™ Plus膜的速度 |
---|
Products
标准宽度(20mm和25mm)和定制小卷的Hi-Flow™ Plus膜也可线下订购母卷膜。每个母卷宽30.5 cm,长100 m。为方便使用,配送前母卷可切割成所需宽度的多个即用型小卷。每个母卷订单收到的小卷数与所需的小卷宽度相关。例如,订购1卷小卷宽度18mm的母卷膜将得到16卷小卷膜。
请联系您当地的客户经理或我们的客服团队,获取自选膜和小卷宽度对应的货号。
生产用Hi-Flow™ Plus膜 - 小卷宽度 |
---|
Hi-Flow™ Plus膜卡标准尺寸
注:另可定制膜卡尺寸。
Hi-Flow™ Plus膜和SureWick®结合垫、样品垫和吸收垫为您的侧向层析检测提供完整的解决方案。
相关资源
- Press Release: Merck Announces the Investment of More Than €440 Million and Expansion of Membrane and Filtration Manufacturing in Ireland
The investment, which is the largest in a single site ever for the Life Science business sector, significantly increases membrane manufacturing capacities and enables the construction of a new filtration manufacturing facility.
- Press Release: Merck Announces $136.7 Million U.S. Government Contract Award for New Lateral Flow Membrane Production Facility in Sheboygan, Wisconsin
Merck has received a $136.7M contract to build a lateral flow membrane facility in Sheboygan, WI, enhancing pandemic preparedness supply.
- Tech Article: IVD Lateral Flow – Sample, Conjugate and Absorbent Pad Basics
选择适宜的侧向层析垫材料是确保检测成功的关键。程序垫由可用于样品垫、结合垫和吸附垫的多孔基质制成。
- Application Note: Bed Volumes of SureWick® Pad Materials
SureWick® pad materials, made from glass fibers and cellulose, are key for lateral flow test strips, influencing sample volume and functionality.
- Brochure: Hi-Flow™ Plus Membranes and SureWick® Pad Materials
We provide materials, components, and custom services, including preassembled membrane cards, to enhance rapid diagnostic test manufacturing efficiency.
- Brochure: Complete Solutions for Lateral Flow Assay Development
Simplify assay development and manufacture with a partner who understands the IVD market
- Brochure: Rapid Lateral Flow Test Strips
感谢您对我们的侧向层析产品和服务感兴趣。烦请填写下表,我们会尽快与您取得联系。
在本期博客中,Shawn Gaskell 将探讨用于新冠病毒和妊娠测试的毛细流膜、其发展历程以及在医学领域的前景。敬请收听,获取更多不凡见解!
2022 年RSV住院人数激增——尤其是在幼儿群体中。专家认为,这种增长与疫情相关行为对免疫力的影响有关。疫苗虽有望治愈疾病,但早期发现和及时治疗至关重要。