主要文件
M9785
Millipore
Ultrafree-MC microcentrifuge filters
working volume 500 μL, Durapore PVDF membrane, pore size 0.65 μm
登录查看公司和协议定价
About This Item
UNSPSC代码:
41103900
NACRES:
NB.24
物料
Durapore PVDF membrane
polypropylene housing
无菌性
non-sterile
制造商/商品名称
Millipore UFC30DV00
参数
12,000 × g max. RCF
工作体积
500 μL
孔径
0.65 μm
正在寻找类似产品? 访问 产品对比指南
一般描述
Units consist of microcentrifuge tube with hinged cap and filter insert. The Ultrafree centrifugal filter units are are well suited for the removal of particles & precipitates from aqueous & some solvent-based samples.
相容性
Use in any standard fixed-angle microcentrifuge rotor.
历史批次信息供参考:
分析证书(COA)