Key Documents
EP022638483
Rubber Mat for Eppendorf® 5804/5804R & 5810/5810R Centrifuges
for 250 mL bucket adapter, pack of 4
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41103911
包裝
pack of 4
製造商/商標名
Eppendorf® 022638483
技術
centrifugation: suitable
相容性
for use with 250 mL rectangular buckets
法律資訊
Eppendorf is a registered trademark of Eppendorf AG