CLS431355
1 of 10
CLS4365
Corning® 聚苯乙烯泡沫塑料管架(50 mL 离心管)
Sigma-Aldrich
R5651
Four-way microtube rack
CLS4366
Corning®通用管架,用于15 mL和50 mL锥形底离心管
R7776
Floating tube rack
HS29025G
Microtube Rack
BR780605
BRAND®微量离心管架
R8509
Scienceware® centrifuge tube rack, epoxy-coated steel
C8358
Cube Rack
Millipore
22092
胰蛋白胨大豆肉汤培养基 - 脱水培养基
R8384
foamed PE
Holds 15 mL tubes
pack of 20 ea
Corning 431355
available only in South Korea
Corning®
实验室器材
实验室容器与储存
实验室耗材合作伙伴
样品处理