主要文件
20943
重复分液器
for use with Hamilton all 10 μL-2.5 mL Hamilton Syringes, for use with Hamilton PB600-1
登录查看公司和协议定价
About This Item
UNSPSC代码:
41122001
制造商/商品名称
Hamilton 83700
包装
1 ea of
相容性
for use with Hamilton PB600-1
for use with Hamilton all 10 μL-2.5 mL Hamilton Syringes
一般描述
精确分配样品。每按一次按钮,即排出注射器容量的 1/50 的样品溶液。适用于 10μL 至 2.5mL 的
Hamilton 注射器。请单独订购注射器。
应用
Repeating Dispenser has been used for catalytic determination of enzymes and metal ions using either closed-loop or continuous-flow analyzers.
历史批次信息供参考:
分析证书(COA)
访问文档库
Catalytic determinations of enzymes and metal ions by sample injection in closed-loop flow systems.
Ramasamy, S. M., A. Iob, and Horacio A. Mottola.
Analytical Chemistry, 51.11, 1637-1639 (1979)