主要文件
TQNXA4RP2
Millipore
Steritest® Equinox Advanced Requalification Protocol
A4 format
登录查看公司和协议定价
About This Item
UNSPSC代码:
41116500
NACRES:
NB.79
相容性
for use with Steritest® Equinox pump.
一般描述
Steritest® Equinox Advanced Requalification protocol is designed as a ready to use guide to perform the requalification of the Steritest® Equinox pump. The requalification protocol can be used to re-certify consistent performance of the pumps upon encountering any change or at regular intervals. On top of some IQ and OQ tests it also covers the periodic review of operator training, SOPs, and OOS results.
应用
Steritest® Equinox Advanced Requalification protocol is used for the re-qualification of Steritest® Equinox pump which is used to perform sterility testing for pharmaceutical products.
法律信息
STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany