包裝
pkg of 1 unit
環保替代產品特色
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
相容性
for use with Milli-Q® SQ 2Series
環保替代類別
一般說明
SQPAK™ RO反渗透滤芯以高达42 L/h流速产出RO纯水。
滤芯′RO膜有效去除99%的水体污染物,包括细菌、离子、颗粒和有机物。
用于安装到Milli-Q® SQ 2系列配置的制水站上,包括Milli-Q® SR 240/240L/240XL和Milli-Q® SQ 240C/240CV系统。
特點和優勢
- 高效去除进水中99%的离子、细菌、颗粒和有机物
- 旋转&锁紧特点使更换轻而易举
- 水机上的指示灯会闪橙色提醒换柱需要
- SQPAK™ RO滤芯的纯水标签和绿色色块同样出现在制水站的交界面,用于提示其纯水作用并指引换新
法律資訊
实验方案
View instructional videos and download User Manuals for Milli-Q® SQ 2Series lab water systems. You’ll be up and running in no time. Buy online systems, accessories, cartridges and services.
View instructional videos and download User Manuals for Milli-Q® SQ 2Series lab water systems. You’ll be up and running in no time. Buy online systems, accessories, cartridges and services.
观看教学视频并下载 Milli-Q® SQ 2 系列实验室纯水系统用户手册。您将快速掌握,并立即投入使用。在线购买系统、配件、纯化柱和服务。
View instructional videos and download User Manuals for Milli-Q® SQ 2Series lab water systems. You’ll be up and running in no time. Buy online systems, accessories, cartridges and services.
相关内容
探索全新的超纯水获取体验现在,任何带有插座的实验台均能获取新鲜的 18.2 MΩ.cm 超纯水。品质稳健操作简单外形紧凑支持扩展绿色替代产品线上购买
Discover a new way to think about ultrapure water. Now any lab bench with a plug can freshly dispense 18.2 MΩ∙cm ultrapure water. Robust Quality. Simple. Compact. Scalable. A Greener Alternative Product. Buy Online.
Discover a new way to think about ultrapure water. Now any lab bench with a plug can freshly dispense 18.2 MΩ∙cm ultrapure water. Robust Quality. Simple. Compact. Scalable. A Greener Alternative Product. Buy Online.
Discover a new way to think about ultrapure water. Now any lab bench with a plug can freshly dispense 18.2 MΩ∙cm ultrapure water. Robust Quality. Simple. Compact. Scalable. A Greener Alternative Product. Buy Online.