跳转至内容
Merck
所有图片(1)

主要文件

203709

Sigma-Aldrich

Insulin Expression Inducer, BRD7389

A cell-permeable naphthoquinolinedione compound that specifically induces insulin secretion in mouse αTC1-cells and promotes transdifferentiation to a β-cell-like status.

别名:

Insulin Expression Inducer, BRD7389, 1-((2-Phenylethyl)amino)-3H-naphtho[1,2,3-de]quinoline-2,7-dione, Insulin Receptor-Related Protein Inhibitor, IRR Inhibitor, INSRR Inhibitor, RSK Inhibitor III

登录查看公司和协议定价


About This Item

经验公式(希尔记法):
C24H18N2O2
分子量:
366.41
MDL编号:
UNSPSC代码:
12352200
NACRES:
NA.51
价格与库存信息目前不能提供

质量水平

方案

≥98% (HPLC)

表单

solid

制造商/商品名称

Calbiochem®

储存条件

OK to freeze
protect from light

颜色

orange-yellow

溶解性

DMSO: 100 mg/mL

运输

ambient

储存温度

2-8°C

SMILES字符串

[nH]1c2c3c(c5c([c](c3ccc2)=O)cccc5)c([c]1=O)NCCc4ccccc4

InChI

1S/C24H18N2O2/c27-23-17-10-5-4-9-16(17)21-20-18(23)11-6-12-19(20)26-24(28)22(21)25-14-13-15-7-2-1-3-8-15/h1-12,25H,13-14H2,(H,26,28)

InChI key

XASCINRGTHLHGM-UHFFFAOYSA-N

一般描述

A cell-permeable naphthoquinolinedione compound that is reported to preferentially inhibit 23 kinases (IC50 ≤10 µM) from a panel of 219, notably IRR (h), CLK2(h), Rsk1(h), Flt3(h), Rsk1(r), Rsk2(h), Haspin(h), Rsk4(h), Melk(h), Drak1(h), Rsk3(h), and CK13(h) (% inhibition by 10 µM BRD7389 = 91, 87, 86, 79, 78, 76, 74, 73, 73, 72, 71, and 71, respectively). Shown to induce β-cell-like morphology in cultured murine pancreatic α-cell αTC1 and stimulate the expression of insulin mRNA (by ~50-fold; 0.85 µM for 5 d) and protein (by 1.5-fold; 3.4 µM for 5 d), as well as other β-cell mRNAs, including Iapp, Npy, Pdx1, and Pax4, in αTC1 cultures. In human primary islets cultures, BRD7389 treatments result in a decrease of overall cell counts over time due to apoptosis induction, while insulin-positive population, and in some cases also glucagon-positive cell population, remains more or less unaffected.

包装

Packaged under inert gas

警告

Toxicity: Regulatory Review (Z)

重悬

Following recontitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

其他说明

Fomina-Yadlin, D., et al. 2010. Proc. Natl. Acad. Sci. USA107, 15099.

法律信息

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

储存分类代码

11 - Combustible Solids

WGK

WGK 3

闪点(°F)

Not applicable

闪点(°C)

Not applicable


分析证书(COA)

输入产品批号来搜索 分析证书(COA) 。批号可以在产品标签上"批“ (Lot或Batch)字后找到。

已有该产品?

在文件库中查找您最近购买产品的文档。

访问文档库

Questions

Reviews

No rating value

Active Filters

我们的科学家团队拥有各种研究领域经验,包括生命科学、材料科学、化学合成、色谱、分析及许多其他领域.

联系技术服务部门