1.50178
1 of 10
Supelco
1.50734
LiChrospher® 100 RP-18 封端 (5 µm) LiChroCART™ 125-4
1.51486
manu-CART™ NT卡套,用于LiChroCART™ 2,3,4,4.6 mm i.d.HPLC 柱
1.50943
LiChrospher® 100 RP-18 (5 µm) LiChroCART™ 125-4
1.52037
Chromolith® Guard Cartridge Holder
1.09535
pH指示条pH 0-14通用指示条
1.50995
LiChrospher® 100 RP-18 封端 (5 µm) LiChroCART™ 250-4
1.51419
manu-CART® "10"-II cartridge holder for LiChroCART® 10 mm i.d. HPLC-cartridges
1.50936
LiChrospher® 100 RP-18 封端 (5 µm) LiChroCART™ 25-4
1.18320
Empty cells
1.50957
LiChrospher® 100 RP-18 (5 µm) LiChroCART™ 4-4
100