主要文件
Z743188
Julabo® bath covers
with built in heat exchanger to fit CORIO CD-BC12
登录查看公司和协议定价
About This Item
UNSPSC代码:
41103706
NACRES:
NB.77
包装
pack of 1 ea
制造商/商品名称
Julabo 9970242
正在寻找类似产品? 访问 产品对比指南
相关类别
一般描述
Bath cover with built-in heat exchanger. To use the heat exchanger, a hose with an inner diameter of 10/12 mm is required.
法律信息
Julabo is a registered trademark of Julabo USA, Inc.
历史批次信息供参考:
分析证书(COA)