Key Documents
CLS2560200
Pyrex® jacketed Graham condenser with drip-tip
jacket L 200 mm, joint: ST/NS 19/38
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41121800
NACRES:
NB.43
接合處
joint: ST/NS19/38
製造商/商標名
Corning 2560-200
護套長度
200 mm
法律資訊
PYREX is a registered trademark of Corning, Inc.