Key Documents
Z515949
Sigma-Aldrich
Aldrich® solvent storage flask
capacity 500 mL, joint: ST/NS 24/40
别名:
bottom flask, flask round, round flask
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41121804
NACRES:
NB.43
材料
borosilicate glass
round bottom
single-neck
接合處
joint: ST/NS24/40
無菌
non-sterile
特點
closure type Standard Taper Joint
包裝
pack of 1
製造商/商標名
Sigma-Aldrich
容量
500 mL
正在寻找类似产品? 访问 产品对比指南
相关类别
一般說明
With ground-glass joint and glass stopcock. Use septum Z565695 or Z565679 (see Septa Section).
法律資訊
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC