主要文件
Z513377
Sigma-Aldrich
Micro spatulas
flat rounded/tapered ends, L 12 in. (305 mm)
登录查看公司和协议定价
About This Item
UNSPSC代码:
41122403
NACRES:
NB.35
物料
nickel-stainless steel
包装
pkg of 6 ea
长度
12 in. (305 mm)
正在寻找类似产品? 访问 产品对比指南
一般描述
Nickel stainless spatula with one rounded and one tapered end. Rounded end approximately 5/16 in. (8 mm) wide x 2 in. (51 mm) long. Tapered end approximately 1/4 in. (6 mm) wide x 1 3/4 in. (44 mm) long.
历史批次信息供参考:
分析证书(COA)