主要文件
Z257400
Spirit-filled low-temperature thermometer
L 305 mm (30.5 cm), temp. range -100/50 °C
登录查看公司和协议定价
About This Item
UNSPSC代码:
41112303
NACRES:
NB.79
价格与库存信息目前不能提供
包装
pack of 1 ea
制造商/商品名称
HB Instruments B60760-1400
参数
-100/50 °C temp. range
长度
305 mm (30.5 cm)
正在寻找类似产品? 访问 产品对比指南
一般描述
These thermometers contain an organic liquid except Z257419 which is filled with a clear Toluene liquid. Total-immersion. 1° divisions for use in general-laboratory, low-temperature applications.
相关产品
产品编号
说明
价格
历史批次信息供参考:
分析证书(COA)
访问文档库
Questions
  1. the description says the thermometer is filled with organic liquid, but doesn't specify what liquid that is. Do you have that information or a safety sheet for the liquid? I'd just like to know how toxic the substance is, e.g. if it contains toluene
    1 answer
    1. This item is a product of Bel-Art. The thermometer contains toluene, as indicated on the manufacturer's Safety Data Sheet (SDS). Please see the link below to access the SDS:
      https://www.belart.com/media/catalogstudio/SDS/Toluene.pdf