主要文件
Z182184
Perfektum® three-way compression-nut stopcock
male Luer lock to male Luer lock to right-side female Luer
登录查看公司和协议定价
About This Item
UNSPSC代码:
41122000
NACRES:
NB.03
特点
male Luer lock to male Luer lock to right-side female Luer
制造商/商品名称
Cadence Science 6014
正在寻找类似产品? 访问 产品对比指南
一般描述
Water tested at 80 psi.
法律信息
Perfektum is a registered trademark of Cadence Science
历史批次信息供参考:
分析证书(COA)