主要文件
SYNF311250
Synthware recovery flask
250 mL, joint: ST/NS 14/20
登录查看公司和协议定价
About This Item
UNSPSC代码:
41121804
NACRES:
NB.51
物料
borosilicate glass flask
pear-shaped
single-neck
接合处
joint: ST/NS14/20
包装
pkg of 1 ea
制造商/商品名称
Synthware F311250
容量
250 mL
250 mL
正在寻找类似产品? 访问 产品对比指南
一般描述
Heavy wall design. All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.
法律信息
Synthware is a trademark of Kemtech America Inc.
历史批次信息供参考:
分析证书(COA)