跳转至内容
Merck

PH005558

Sigma-Aldrich

N-Ethyl-N′-((2Z)-5-nitro-3-(2-propynyl)-1,3-thiazol-2(3H)-ylidene)urea

AldrichCPR

登录查看公司和协议定价


About This Item

线性分子式:
C9H10N4O3S
CAS号:
价格与库存信息目前不能提供

SMILES字符串

O=C(NCC)/N=C1N(CC#C)C=C([N+]([O-])=O)S/1

InChI

1S/C9H10N4O3S/c1-3-5-12-6-7(13(15)16)17-9(12)11-8(14)10-4-2/h1,6H,4-5H2,2H3,(H,10,14)/b11-9-

其他说明

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

闪点(°F)

Not applicable

闪点(°C)

Not applicable


历史批次信息供参考:

分析证书(COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our 文件 section.

如需帮助,请联系 客户支持

已有该产品?

在文件库中查找您最近购买产品的文档。

访问文档库

Questions

Reviews

No rating value

Active Filters

我们的科学家团队拥有各种研究领域经验,包括生命科学、材料科学、化学合成、色谱、分析及许多其他领域.

联系技术服务部门