主要文件
BR13948
BRAND®备用滴定管延长管,适用于紧凑型滴定管
size 50 mL, BLAUBRAND®, clear borosilicate glass 3.3
别名:
BLAUBRAND®, 滴定管, 紧凑型滴定管, 配件
登录查看公司和协议定价
About This Item
UNSPSC代码:
41121808
NACRES:
NB.43
物料
clear borosilicate glass 3.3
包装
pack of 1 ea
制造商/商品名称
BRAND 13948
长度
705 mm
尺寸
50 mL
正在寻找类似产品? 访问 产品对比指南
一般描述
The BRAND® spare burette tubes for compact burettes are crafted from high-quality Boro 3.3 clear glass, ensuring both durability and visibility for accurate liquid measurement. These spare tubes are specifically designed to be compatible with BLAUBRAND® and SILBERBRAND compact burettes, making them a reliable choice for maintaining your liquid handling systems.
法律信息
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
历史批次信息供参考:
分析证书(COA)