Key Documents
BR13928
BRAND®紧凑型SILBERBRAND滴定管,带PTFE旋塞
volume 50 mL
别名:
SILBERBRAND, 滴定管, 紧凑型滴定管
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41121808
NACRES:
NB.43
材料
borosilicate glass (5.4)
製造商/商標名
BRAND 13928
體積測量範圍
50 mL, accuracy: ±0.10 mL
容積
50 mL
正在寻找类似产品? 访问 产品对比指南
一般說明
The New Generation of Burettes from BRAND. With PTFE stopcock, fast to dismantle and easy to clean. Simple repair - all individual components replaceable. Borosilicate glass 3.3, removable PTFE stopcock. 25 ml: Error limits according to class B, DIN EN ISO 385. 50 ml: To DIN EN ISO 385, class B.
法律資訊
BRAND is a registered trademark of BRAND GMBH + CO KG