主要文件
925470
Sigma-Aldrich
Calibrated ChemBeads Scoop
登录查看公司和协议定价
About This Item
UNSPSC代码:
12352101
NACRES:
NA.52
一般描述
Calibrated ChemBeads scoops allow you to weigh ChemBeads products volumetrically. These scoops have been designed to consistently deliver the calibrated amount of ChemBeads quickly and efficiently. The scoops are made of a chemical resistant plastic, acrylonitrile butadiene styrene, and can be reused multiple times, however they should not be cleaned with strong solvents.
There are three sizes of Calibrated ChemBeads Scoops available or a kit containing a set of all three. The sizes are outlined below with Catalog Number and Calibrated Weight:
925470-5MG 5 milligrams
925470-10MG 10 milligrams
925470-50MG 50 milligrams
925470-1SET one of each scoop calibrated to deliver 5, 10, or 50 milligrams
ChemBeads are chemical coated glass beads. ChemBeads offer improved flowability and chemical uniformity perfect for automated solid dispensing and high-throughput experimentation. The method of creating ChemBeads uses no other chemicals or surfactants allowing the user to accurately dispense sub-milligram amounts of chemical.
Learn more about ChemBeads products
历史批次信息供参考:
分析证书(COA)
访问文档库
商品
ChemBeads, catalyst-coated glass beads, dispense solid chemical reagents for nanomole-scale high-throughput reaction screening.
ChemBeads, catalyst-coated glass beads, dispense solid chemical reagents for nanomole-scale high-throughput reaction screening.
ChemBeads, catalyst-coated glass beads, dispense solid chemical reagents for nanomole-scale high-throughput reaction screening.
ChemBeads, catalyst-coated glass beads, dispense solid chemical reagents for nanomole-scale high-throughput reaction screening.