Z379298
Deepwell 96 well plate
Cover only
Synonym(s):
plate lid, plate cover
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122107
NACRES:
NB.22
material:
polypropylene
size:
96 wells
well volume:
2 mL
sterility:
non-sterile
Recommended Products
material
polypropylene
sterility
non-sterile
packaging
pack of 24 ea
manufacturer/tradename
Bel-Art F37860-0004
size
96 wells
well volume
2 mL
color
none
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
These high-form polypropylene 96 well plates expand the flexibility of multichannel or automated pipetting systems. Use for non-binding protein assays, enzyme or receptor assays, or with cell harvesters. The 2 mL per well capacity makes it possible to do serial dilution procedures that are impossible with normal multiwell plates.
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service