Skip to Content
Merck
All Photos(1)

Key Documents

97088

Supelco

2-Aminobenzamide

analytical standard

Synonym(s):

Anthranilamide, 2-AB, 2-Aminobenzamide, Anthranilic acid amide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(H2N)C6H4CONH2
CAS Number:
Molecular Weight:
136.15
Beilstein:
508509
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.21

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

technique(s)

HPLC: suitable

mp

111-113 °C (lit.)

format

neat

storage temp.

2-30°C

SMILES string

NC(=O)c1ccccc1N

InChI

1S/C7H8N2O/c8-6-4-2-1-3-5(6)7(9)10/h1-4H,8H2,(H2,9,10)

InChI key

PXBFMLJZNCDSMP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used for non-selective, efficient fluorescent labeling of glycans. Slightly less sensitive than anthranilic acid (2-AA) for glycan labeling.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

>365.0 °F

Flash Point(C)

> 185 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service