Key Documents
MTCP3802QW
MTC Bio Pure.AmpqPCR 8 Tube Strips
capacity 0.2 mL, white tube, clear cap (optical), flat cap
Synonym(s):
PCR sample storage tubes, plastic tube, sample storage tube
About This Item
UNSPSC Code:
41121800
NACRES:
NB.15
material
clear cap (optical)
clear tube
flat cap
polypropylene
white tube
sterility
non-sterile
packaging
pack of 120 ea
manufacturer/tradename
MTC Bio P3802-QW
capacity
0.2 mL
General description
0.2ml qPCR 8-Strip (With SEPARATE Optical Strip Caps), WHITE, 120/pk
Pure.Amp 0.1mL qPCR strips strips are precision manufactured to maintain tight dimensional tolerances of ultra-thin wall thickness, for improved thermal transfer and are certified free of RNase, DNase, DNA and PCR inhibitors, and are made of heat-resistant medical-grade polypropylene.
Legal Information
MTC is a trademark of MTC Bio
Pure.Amp is a trademark of MTC Bio
Choose from one of the most recent versions:
Certificates of Analysis (COA)