Key Documents
MTCR4015
MTC Bio CoolCaddy Cold Station
to hold, PCR plate, tubes and cryovials
Synonym(s):
PCR cooler, benchtop cooler, sample tube
About This Item
UNSPSC Code:
41103037
NACRES:
NA.21
material
acrylonitrile-butadiene-styrene (ABS)
feature
to hold PCR plate, tubes and cryovials
packaging
pack of 1 ea
manufacturer/tradename
MTC Bio R4015
General description
CoolCaddy keeps your reagents and samples cool on the lab bench without using crushed ice. Simply store your CoolCaddy overnight in any standard freezer and it’s ready to hold temperatures between 2° and 4°C for up to 5 hours. Tubes, plates, strips and multi-channel reservoir solutions can all be kept cool while you perform your experiments.
Features and Benefits
  • Maintains 2-4°C for up to 5 hours
  • Keeps samples cool, but not frozen
  • Accommodates several different vessel types
  • 1.5/2.0mL Microtubes: 9 Tubes
  • 2.0mL Cryotubes: 8 Tubes
  • 0.5mL Microtubes: 5 Tubes
  • PCR Plates: 1 Plate
  • PCR Tubes: 96 Tubes or 12 Strips
  • Reservoirs: 1 x 50mL Reservoir
Legal Information
CoolCaddy is a trademark of MTC Bio
MTC is a trademark of MTC Bio
Choose from one of the most recent versions:
Certificates of Analysis (COA)