Skip to Content
Merck
All Photos(1)

Key Documents

1B8-F11

Reactivity: Anti-Bovine Herpes Virus 1, 95022202

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Mouse x Mouse Hybridoma

ImmunoDon

BALB/c mouse spleen

Myeloma

Sp2/0

Reactivity

Anti-Bovine Herpes Virus 1

technique(s)

cell culture | mammalian: suitable

isotype

IgG1

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse Sp2/0 x Mouse BALB/c spleen

Cell Line Description

The antibody is directed against an immunodominant glycoprotein of bovine herpes virus 1, BHV-1, (infectious rhinotracheitis virus). Virus neutralisation could be observed with ascites fluids prepared in BALB/c mice.

Immunogen

Bovine herpes virus 1

Application

In vitro potency testing of BHV-1 vaccines.

Culture Medium

DMEM + 2mM glutamine + 2 5% Foetal Bovine Serum (FBS) + 5% Horse Serum (HS)

Subculture Routine

Maintain cultures between 100,000 - 1,000,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service