Skip to Content
Merck
All Photos(1)

Key Documents

BC9

Reactivity: Anti-TNF, bovine, 95030301

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Mouse x Mouse Hybridoma

ImmunoDon

BALB/c mouse spleen

Myeloma

P3X63Ag8.653

Reactivity

Anti-TNF, bovine

technique(s)

cell culture | mammalian: suitable

isotype

IgG1

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse P3X63Ag8.653 x Mouse BALB/c spleen

Cell Line Description

The monoclonal antibody BC9 detects bovine tumour necrosis factor (TNF) in a radioimmunoassay with a detection limit of 24 pg/ml. Bovine recombinant TNF can be neutralised in vitro by addition of BC9. Cytotoxic assays showed a neutralising effect using L929 and WEHI 164 clone 13 cells.

Immunogen

Bovine recombinant TNF-alpha

Application

Detection of TNF in bioassay or radioimmunoassay, in vivo studies (neutralisation of TNF).

Culture Medium

RPMI 1640 + 2mM glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 5-6x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service